Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RALBP1 associated Eps domain containing 2 | REPS2 | POB1 | Anti-sense | Chr. X - 16946691 - 17194274 on NCBI Build GRCh38 | |
| Downstream | NHS actin remodeling regulator | NHS |
CTRCT40, CXN, SCML1 |
248.4kb | Anti-sense | Chr. X - 17375420 - 17735994 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map