Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
FERM, ARH/RhoGEF and pleckstrin domain protein 1 |
FARP1 |
CDEP, FARP1-IT1, PLEKHC2, PPP1R75 |
-7.7kb | Anti-sense | Chr. 13 - 98142562 - 98449773 on NCBI Build GRCh38 |
| Downstream | STK24 antisense RNA 1 | STK24-AS1 | 118.1kb | Anti-sense | Chr. 13 - 98577585 - 98578842 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map