Assay Details
| Gene Symbol: ING3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019071.2 | NP_061944.2 | 7 - 8 | 690 | 77 | |
| GenBank mRNA | AF074968.1 | - | 7 - 8 | 667 | 77 | |
| AK000096.1 | - | 7 - 8 | 691 | 77 | ||
| AK293682.1 | - | 6 - 7 | 576 | 77 | ||
| AL603623.1 | - | 7 - 8 | 667 | 77 | ||
| AY007790.1 | - | 7 - 8 | 671 | 77 | ||
| BC010851.1 | - | 6 - 7 | 540 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000734.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
regulation of growth
positive regulation of apoptotic process
histone H4 acetylation
histone H2A acetylation
histone acetyltransferase activity
zinc ion binding
methylated histone binding
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
regulation of growth
positive regulation of apoptotic process
histone H4 acetylation
histone H2A acetylation
histone acetyltransferase activity
zinc ion binding
methylated histone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07011716.hg.1 | TC07000734.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07011716.hg.1 | ENST00000315870.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07011716.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07011716.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700163334.hg.1 | TC0700008926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000315870.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000339121.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000427726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000431467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000445699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | ENST00000497502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_019071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008926.hg.1 | NM_198267.1 | Refseq |

ING3
Genomic Map