Assay Details
| Gene Symbol: POT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516006.1 | XP_011514308.1 | 4 - 5 | 563 | 155 | |
| XM_011516007.1 | XP_011514309.1 | 4 - 5 | 562 | 155 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011516006.1, XM_011516007.1 transcripts.
Set Membership: |
|

POT1
Genomic Map