Assay Details
| Gene Symbol: POT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042594.1 | NP_001036059.1 | 4 - 5 | 565 | 123 | |
| NM_015450.2 | NP_056265.2 | 4 - 5 | 565 | 123 | ||
| NR_003102.1 | 4 - 5 | 565 | 123 | |||
| NR_003103.1 | 4 - 5 | 565 | 123 | |||
| NR_003104.1 | 4 - 5 | 565 | 123 | |||
| XM_006715917.3 | XP_006715980.1 | 4 - 5 | 555 | 123 | ||
| XM_017011941.1 | XP_016867430.1 | 4 - 5 | 563 | 123 | ||
| XM_017011942.1 | XP_016867431.1 | 4 - 5 | 556 | 123 | ||
| GenBank mRNA | AK001935.1 | - | 1 - 2 | 94 | 123 | |
| AK022580.1 | - | 4 - 5 | 402 | 123 | ||
| AK302263.1 | - | 4 - 5 | 720 | 123 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK001935.1,AK022580.1,AK302263.1,NM_001042594.1,NM_015450.2,XM_006715917.3,XM_017011941.1,XM_017011942.1 transcripts.
Set Membership: |
|

POT1
Genomic Map