Assay Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI479646.1 | - | 2 - 3 | 362 | 123 | |
| BC002779.2 | - | 2 - 3 | 369 | 123 | ||
| BC094884.1 | - | 2 - 3 | 369 | 123 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000411000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000299314.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000392919.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000411000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000549940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000550718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | ENST00000552681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011711.hg.1 | NM_024312.4 | Refseq |

GNPTAB
Genomic Map