Assay Details
| Gene Symbol: SDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006843.2 | NP_006834.2 | 4 - 5 | 450 | 62 | |
| GenBank mRNA | AK292760.1 | - | 4 - 5 | 446 | 62 | |
| BC020750.1 | - | 4 - 5 | 449 | 4662951 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001989.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12026137.hg.1 | ENST00000257549.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026137.hg.1 | ENST00000446302.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026137.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026137.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026137.hg.1 | TC12001989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000546785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000552280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011994.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000257549.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000547342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | ENST00000553112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | NM_006843.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200193437.hg.1 | TC1200011994.hg.1 | Clariom D TC |

SDS
Genomic Map