Assay Details
| Gene Symbol: SLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK297452.1 | - | 4 - 5 | 512 | 153 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000265140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000450932.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000466957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000475916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000493934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000504099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | ENST00000508383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008107.hg.1 | NM_032290.3 | Refseq |

SLF1
Genomic Map