Assay Details
| Gene Symbol: HRH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF321912.1 | - | 3 - 4 | 825 | 165 | |
| AF346904.1 | - | 3 - 4 | 825 | 165 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000317393.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000340177.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | ENST00000611492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009736.hg.1 | NM_007232.2 | Refseq |

HRH3
Genomic Map