Assay Details
| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022095.3 | NP_071378.1 | 25 - 26 | 3875 | 79 | |
| XM_005260504.4 | XP_005260561.1 | 25 - 26 | 4588 | 79 | ||
| XM_017028012.1 | XP_016883501.1 | 24 - 25 | 3646 | 79 | ||
| GenBank mRNA | AF395833.1 | - | 25 - 26 | 3852 | 79 | |
| AK026157.1 | - | 17 - 18 | 2390 | 79 | ||
| AK074214.1 | - | 10 - 11 | 1510 | 79 | ||
| AK122963.1 | - | 8 - 9 | 2066 | 79 | ||
| AK296989.1 | - | 22 - 23 | 3446 | 79 | ||
| BC041331.2 | - | 22 - 23 | 3090 | 79 | ||
| BC142981.1 | - | 22 - 23 | 3090 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000894.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012199.hg.1 | ENST00000243961.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012199.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012199.hg.1 | ENST00000426788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012199.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012199.hg.1 | TC20000894.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000164774.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164774.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164774.hg.1 | TC2000009253.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164774.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164774.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |

ZNF335
Genomic Map