Assay Details
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022095.3 | NP_071378.1 | 8 - 9 | 1474 | 62 | |
XM_005260504.4 | XP_005260561.1 | 8 - 9 | 2187 | 62 | ||
XM_011528979.2 | XP_011527281.1 | 7 - 8 | 1842 | 62 | ||
XM_017028012.1 | XP_016883501.1 | 7 - 8 | 1245 | 62 | ||
GenBank mRNA | AF395833.1 | - | 8 - 9 | 1451 | 62 | |
AK296989.1 | - | 5 - 6 | 1045 | 62 | ||
BC041331.2 | - | 5 - 6 | 689 | 62 | ||
BC142981.1 | - | 5 - 6 | 689 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000894.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20012218.hg.1 | ENST00000243961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012218.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012218.hg.1 | ENST00000426788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012218.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20012218.hg.1 | TC20000894.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | NM_022095.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | TC2000009253.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | ENST00000322927.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | ENST00000475002.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164792.hg.1 | NM_022095.3 | Refseq |
