Assay Details
| Gene Symbol: SERINC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006811.3 | NP_006802.1 | 6 - 7 | 949 | 89 | |
| NM_198941.2 | NP_945179.1 | 6 - 7 | 949 | 89 | ||
| XM_017027601.1 | XP_016883090.1 | 6 - 7 | 843 | 89 | ||
| GenBank mRNA | AF112227.1 | - | 6 - 7 | 861 | 89 | |
| AF153979.1 | - | 6 - 7 | 878 | 89 | ||
| AK222923.1 | - | 6 - 7 | 871 | 89 | ||
| AK295120.1 | - | 7 - 8 | 824 | 89 | ||
| AK300618.1 | - | 7 - 8 | 946 | 89 | ||
| AK309993.1 | - | 7 - 8 | 1008 | 89 | ||
| BC006088.1 | - | 6 - 7 | 854 | 3641108 | 89 | |
| U49188.1 | - | 6 - 7 | 890 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000867.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylserine metabolic process
sphingolipid metabolic process
detection of virus
L-serine transport
innate immune response
defense response to virus
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
L-serine transmembrane transporter activity
phosphatidylserine metabolic process
sphingolipid metabolic process
detection of virus
L-serine transport
innate immune response
defense response to virus
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
L-serine transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | ENST00000255175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | ENST00000538937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | ENST00000541235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011824.hg.1 | TC20000867.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | TC2000009196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164272.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000255175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000342374.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000411544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | ENST00000468234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_006811.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009196.hg.1 | NM_198941.2 | Refseq |

SERINC3
Genomic Map