Assay Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181805.2 | NP_861521.1 | 2 - 3 | 348 | 70 | |
| XM_017027614.1 | XP_016883103.1 | 1 - 2 | 227 | 70 | ||
| GenBank mRNA | BG717090.1 | - | 2 - 3 | 262 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000334.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_181805.2,XM_017027614.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004734.hg.1 | ENST00000372891.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004734.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004734.hg.1 | TC20000334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151760.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151760.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151760.hg.1 | TC2000007457.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151760.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151760.hg.1 | NM_181805.2 | Refseq |

PKIG
Genomic Map