Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007238.4 | NP_009169.3 | 2 - 3 | 301 | 61 | |
| GenBank mRNA | AF072864.1 | - | 2 - 3 | 223 | 61 | |
| AK023879.1 | - | 2 - 3 | 219 | 61 | ||
| AK316589.1 | - | 2 - 3 | 281 | 61 | ||
| AL136648.1 | - | 2 - 3 | 272 | 61 | ||
| BC001147.1 | - | 2 - 3 | 202 | 3533572 | 61 | |
| CR533471.1 | - | 2 - 3 | 178 | 61 | ||
| DB495125.1 | - | 2 - 3 | 301 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000774.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20010367.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010367.hg.1 | ENST00000409299.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010367.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010367.hg.1 | TC20000774.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | TC2000008895.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162008.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000217398.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000344022.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | ENST00000409299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_007238.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008895.hg.1 | NM_183397.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 155879 | - | - |

PXMP4
Genomic Map