| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001940.hg.1 |
ENST00000255008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001940.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001940.hg.1 |
TC20000151.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001941.hg.1 |
ENST00000255008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001941.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001941.hg.1 |
TC20000151.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006945.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006945.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000147478.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147478.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000147478.hg.1 |
TC2000006945.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000147478.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147478.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000147479.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147479.hg.1 |
NM_001052.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000147479.hg.1 |
TC2000006945.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000147479.hg.1 |
ENST00000255008.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147479.hg.1 |
NM_001052.2 |
Refseq |