Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015352.1 | NP_056167.1 | 1 - 2 | 172 | 63 | |
| NM_172236.1 | NP_758436.1 | 1 - 2 | 172 | 63 | ||
| GenBank mRNA | AF375884.1 | - | 1 - 2 | 172 | 63 | |
| AK291033.1 | - | 1 - 2 | 179 | 63 | ||
| AL832023.1 | - | 1 - 2 | 104 | 63 | ||
| BC000582.2 | - | 1 - 2 | 172 | 3162533 | 63 | |
| D80002.2 | - | 1 - 2 | 169 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000208.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
somitogenesis
fucose metabolic process
regulation of transcription, DNA-templated
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
embryo development
O-glycan processing
protein O-linked fucosylation
fucosyltransferase activity
peptide-O-fucosyltransferase activity
angiogenesis
somitogenesis
fucose metabolic process
regulation of transcription, DNA-templated
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
embryo development
O-glycan processing
protein O-linked fucosylation
fucosyltransferase activity
peptide-O-fucosyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | ENST00000375749.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | ENST00000539210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002558.hg.1 | TC20000208.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000408740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NR_031726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | ENST00000375749.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | ENST00000486717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | ENST00000539210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002559.hg.1 | TC20000208.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002560.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002560.hg.1 | ENST00000375749.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002560.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002560.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002560.hg.1 | TC20000208.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | TC2000007110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148581.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | TC2000007110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148579.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000408740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NR_031726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | NM_172236.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | TC2000007110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | ENST00000375730.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | ENST00000375749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | NM_015352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148582.hg.1 | NM_172236.1 | Refseq |

POFUT1
Genomic Map