Assay Details
| Gene Symbol: CPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001871.2 | NP_001862.2 | 5 - 6 | 498 | 88 | |
| GenBank mRNA | AJ224866.1 | - | 5 - 6 | 480 | 88 | |
| AK313940.1 | - | 5 - 6 | 486 | 88 | ||
| BC015338.1 | - | 5 - 6 | 479 | 4422620 | 88 | |
| BI770145.1 | - | 5 - 6 | 498 | 88 | ||
| BT009910.1 | - | 5 - 6 | 475 | 88 | ||
| M81057.1 | - | 5 - 6 | 498 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000799.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03014349.hg.1 | TC03000799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014349.hg.1 | ENST00000282957.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014349.hg.1 | ENST00000491148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03014349.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165366.hg.1 | TC0300009120.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000282957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000462345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000465718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000468341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000473621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000476847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000484877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000491148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000494888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | ENST00000498639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009120.hg.1 | NM_001871.2 | Refseq |

CPB1
Genomic Map