Assay Details
| Gene Symbol: KCNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173197.2 | NP_775289.1 | 4 - 5 | 692 | 90 | |
| GenBank mRNA | AF367019.1 | - | 4 - 5 | 340 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000343195.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000348850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000353068.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000355657.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000356640.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000358038.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000359877.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000370046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000370059.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000434163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | ENST00000461105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020968.hg.1 | TC10001614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000343195.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000348850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000353068.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000355657.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000356640.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000358038.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000359877.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000370046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000370059.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000434163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | ENST00000461105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020970.hg.1 | TC10001614.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000239117.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000343195.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000348850.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000353068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000355657.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000356640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000358038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000370046.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000434163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000460388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000461105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000472764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000239117.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000343195.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000348850.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000353068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000355657.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000356640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000358038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000370046.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000434163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000460388.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000461105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | ENST00000472764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179793.hg.1 | TC1000011661.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000239117.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000343195.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000348850.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000353068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000355657.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000356640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000358038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000370046.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000434163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000461105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000472764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000239117.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000343195.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000348850.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000353068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000355657.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000356640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000358038.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000370046.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000434163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000461105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | ENST00000472764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_014591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173191.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173192.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173194.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173195.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | NM_173197.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179794.hg.1 | TC1000011661.hg.1 | Clariom D TC |

KCNIP2
Genomic Map