Assay Details
| Gene Symbol: KCNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539773.1 | XP_011538075.1 | 2 | 619 | 100 | |
| XM_011539774.1 | XP_011538076.1 | 2 | 619 | 100 | ||
| XM_011539775.1 | XP_011538077.1 | 2 | 619 | 100 | ||
| XM_011539776.1 | XP_011538078.1 | 2 | 619 | 100 | ||
| XM_011539779.1 | XP_011538081.1 | 2 | 619 | 100 | ||
| XM_011539781.2 | XP_011538083.1 | 2 | 619 | 100 | ||
| XM_011539782.2 | XP_011538084.1 | 2 | 571 | 100 | ||
| XM_011539784.1 | XP_011538086.1 | 2 | 619 | 100 | ||
| XM_017016207.1 | XP_016871696.1 | 2 | 619 | 100 | ||
| XM_017016208.1 | XP_016871697.1 | 2 | 619 | 100 | ||
| XM_017016209.1 | XP_016871698.1 | 2 | 619 | 100 | ||
| XM_017016210.1 | XP_016871699.1 | 2 | 619 | 100 | ||
| XM_017016211.1 | XP_016871700.1 | 2 | 619 | 100 | ||
| XM_017016212.1 | XP_016871701.1 | 2 | 619 | 100 | ||
| XM_017016213.1 | XP_016871702.1 | 2 | 619 | 100 | ||
| XM_017016214.1 | XP_016871703.1 | 2 | 619 | 100 | ||
| XM_017016216.1 | XP_016871705.1 | 2 | 619 | 100 | ||
| XM_017016217.1 | XP_016871706.1 | 2 | 571 | 100 | ||
| XM_017016219.1 | XP_016871708.1 | 2 | 619 | 100 | ||
| XM_017016220.1 | XP_016871709.1 | 2 | 619 | 100 | ||
| GenBank mRNA | AF349445.1 | - | 1 | 75 | 100 | |
| AK124355.1 | - | 2 | 462 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

KCNMA1
Genomic Map