Assay Details
| Gene Symbol: MND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253861.1 | NP_001240790.1 | 1 - 2 | 88 | 124 | |
| NM_032117.3 | NP_115493.1 | 1 - 2 | 88 | 124 | ||
| NR_045605.1 | 1 - 2 | 88 | 124 | |||
| GenBank mRNA | BM833450.1 | - | 1 - 2 | 88 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000773.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001253861.1,NM_032117.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | TC04000773.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | ENST00000240488.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | ENST00000504860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010617.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | TC04000773.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | ENST00000240488.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010614.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000578990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161995.hg.1 | TC0400009037.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000578990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161991.hg.1 | TC0400009037.hg.1 | Clariom D TC |

MND1
Genomic Map