Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
TC11003466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000357685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000361053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000393032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000438022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000526088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000531169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
ENST00000532593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
NM_001037290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
NM_001256397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
NM_001256398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
NM_001256400.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012612.hg.1 |
NM_031938.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000357685.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000361053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000438022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000460924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000525175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000526088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000531169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001037290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_031938.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000357685.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000361053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000438022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000460924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000525175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000526088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000531169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
ENST00000532716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001037290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_001256400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
NM_031938.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202481.hg.1 |
TC1100013093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000357685.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000361053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000438022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000460924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000494860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000526088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000530677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000531169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000532593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000532716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001037290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_031938.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000357685.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000361053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000438022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000460924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000494860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000526088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000530677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000531169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000532593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
ENST00000532716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001037290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_001256400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
NM_031938.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202477.hg.1 |
TC1100013093.hg.1 |
Clariom D TC |