Assay Details
| Gene Symbol: AKAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016248.3 | NP_057332.1 | 10 - 11 | 5534 | 104 | |
| XM_005266248.3 | XP_005266305.1 | 9 - 10 | 5428 | 104 | ||
| XM_005266249.3 | XP_005266306.1 | 10 - 11 | 5535 | 104 | ||
| XM_011534903.2 | XP_011533205.1 | 10 - 11 | 5531 | 104 | ||
| XM_011534904.1 | XP_011533206.1 | 10 - 11 | 5727 | 104 | ||
| XM_011534905.1 | XP_011533207.1 | 9 - 10 | 5485 | 104 | ||
| XM_011534906.2 | XP_011533208.1 | 9 - 10 | 5703 | 104 | ||
| XM_017020381.1 | XP_016875870.1 | 10 - 11 | 5585 | 104 | ||
| XM_017020382.1 | XP_016875871.1 | 8 - 9 | 5375 | 104 | ||
| XM_017020383.1 | XP_016875872.1 | 8 - 9 | 5563 | 104 | ||
| GenBank mRNA | AB014529.2 | - | 3 - 4 | 3474 | 104 | |
| AF176555.1 | - | 10 - 11 | 5534 | 104 | ||
| BE694434.1 | - | 3 - 4 | 391 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000152.hg.1
Set Membership: |
|

AKAP11
Genomic Map