Assay Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032532.2 | NP_115921.2 | 13 - 14 | 4375 | 130 | |
| XM_011536190.2 | XP_011534492.1 | 12 - 13 | 4298 | 130 | ||
| XM_011536191.2 | XP_011534493.1 | 10 - 11 | 4016 | 130 | ||
| GenBank mRNA | AB058769.2 | - | 11 - 12 | 3861 | 130 | |
| AJ586132.1 | - | 3 - 4 | 295 | 130 | ||
| AK127635.1 | - | 3 - 4 | 383 | 130 | ||
| AL832410.1 | - | 3 - 4 | 592 | 130 | ||
| BC146783.1 | - | 11 - 12 | 3861 | 9007239 | 130 | |
| BC150607.1 | - | 12 - 13 | 4050 | 9056977 | 130 | |
| DQ009660.1 | - | 13 - 14 | 4375 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001142.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ586132.1 transcripts.
Set Membership: |
|

FNDC1
Genomic Map