Assay Details
| Gene Symbol: HMGN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010172.1 | XP_016865661.1 | 2 - 3 | 284 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010172.1 transcripts.
Set Membership: |
|

HMGN4
Genomic Map