Assay Details
| Gene Symbol: LY86-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026970.1 | 1 - 2 | 198 | 139 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lymphocyte antigen 86 | LY86 |
MD-1, MD1, MMD-1, dJ80N2.1 |
Anti-sense | Chr. 6 - 6588639 - 6654983 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928004 | LOC101928004 | 72.1kb | Sense | Chr. 6 - 6694799 - 6801211 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001247.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06014835.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014835.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014835.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014837.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014837.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014832.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014832.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014831.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014831.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014831.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014836.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014836.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000403621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014830.hg.1 | TC06001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014830.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014830.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172756.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172774.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172772.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172776.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172776.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172776.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172764.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172775.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172775.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172775.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172775.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172775.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600172763.hg.1 | TC0600010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000403621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |

LY86-AS1
Genomic Map