Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135218.1 | NP_001128690.1 | 1 - 2 | 434 | 131 | |
| GenBank mRNA | BC011986.1 | - | 1 - 2 | 347 | 3882343 | 131 |
| BE787487.1 | - | 1 - 2 | 326 | 131 | ||
| CX865268.1 | - | 1 - 2 | 356 | 131 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002427.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC011986.1,NM_001135218.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of signal transduction by p53 class mediator
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein heterodimerization activity
DNA-templated transcription, initiation
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of signal transduction by p53 class mediator
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037406.hg.1 | TC01002427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037406.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037406.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037404.hg.1 | TC01002427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037404.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037404.hg.1 | ENST00000373824.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037404.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037404.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200155.hg.1 | TC0100013483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200154.hg.1 | TC0100013483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100200157.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200157.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200157.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100200157.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200157.hg.1 | TC0100013483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000263974.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000373824.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000471683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | ENST00000495422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_001135218.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013483.hg.1 | NM_005644.3 | Refseq |

TAF12
Genomic Map