Assay Details
| Gene Symbol: NPY6R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002713.3 | 1 - 2 | 261 | 148 | ||
| GenBank mRNA | DC357138.1 | - | 1 - 2 | 261 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPY6R| Gene Symbol: NPY6R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002713.3 | 1 - 2 | 261 | 148 | ||
| GenBank mRNA | DC357138.1 | - | 1 - 2 | 261 | 148 | |
Set Membership: |
|