Assay Details
| Gene Symbol: AGBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC030651.2 | - | 12 - 13 | 2209 | 5267615 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165736.hg.1 | TC0700009240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000275763.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000435976.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000436302.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000455283.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000470839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000476700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | ENST00000494702.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009240.hg.1 | NM_178563.3 | Refseq |

AGBL3
Genomic Map