Assay Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281768.1 | NP_001268697.1 | 1 - 2 | 53 | 87 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281768.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
cAMP-mediated signaling
activation of protein kinase A activity
response to drug
regulation of circadian rhythm
cellular response to calcium ion
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
calmodulin binding
ATP binding
calcium- and calmodulin-responsive adenylate cyclase activity
metal ion binding
renal water homeostasis
cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
cAMP-mediated signaling
activation of protein kinase A activity
response to drug
regulation of circadian rhythm
cellular response to calcium ion
cellular response to glucagon stimulus
cellular response to forskolin
adenylate cyclase activity
calmodulin binding
ATP binding
calcium- and calmodulin-responsive adenylate cyclase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000468353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007426.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151665.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151665.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151665.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151665.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151665.hg.1 | TC0700007426.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000297323.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000432715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | ENST00000621543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | NM_001281768.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | NM_021116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700151670.hg.1 | TC0700007426.hg.1 | Clariom D TC |

ADCY1
Genomic Map