Assay Details
| Gene Symbol: CCDC154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143980.1 | NP_001137452.1 | 8 - 9 | 1069 | 140 | |
| GenBank mRNA | JN935901.1 | - | 7 - 8 | 902 | 140 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000752.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16010987.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010987.hg.1 | ENST00000409671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010987.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010987.hg.1 | TC16000752.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010986.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010986.hg.1 | ENST00000409671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010986.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010986.hg.1 | TC16000752.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168430.hg.1 | TC1600009041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | ENST00000389176.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | ENST00000409671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | NM_001143980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600168429.hg.1 | TC1600009041.hg.1 | Clariom D TC |

CCDC154
Genomic Map