Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281450.1 | NP_001268379.1 | 4 - 5 | 930 | 90 | |
XM_011515789.2 | XP_011514091.2 | 4 - 5 | 858 | 90 | ||
GenBank mRNA | AK057366.1 | - | 4 - 5 | 931 | 90 | |
AY081949.1 | - | 4 - 5 | 930 | 90 | ||
BC143787.1 | - | 4 - 5 | 772 | 9052301 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181492.hg.1 | TC0700011489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000488217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | ENST00000493498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011489.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | ENST00000333149.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | ENST00000453152.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_001281450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_001281451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | NM_178125.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700181491.hg.1 | TC0700011489.hg.1 | Clariom D TC |
