Assay Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259671.3 | XP_005259728.1 | 4 - 5 | 896 | 116 | |
| XM_005259672.3 | XP_005259729.1 | 4 - 5 | 897 | 116 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |

MIDN
Genomic Map