Assay Details
| Gene Symbol: RIPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003821.5 | NP_003812.1 | 2 - 3 | 647 | 118 | |
| GenBank mRNA | AF027706.1 | - | 2 - 3 | 557 | 118 | |
| AF064824.1 | - | 2 - 3 | 575 | 118 | ||
| AF078530.1 | - | 2 - 3 | 333 | 118 | ||
| AK075213.1 | - | 2 - 3 | 551 | 118 | ||
| AY358814.1 | - | 2 - 3 | 584 | 118 | ||
| AY524045.1 | - | 2 - 3 | 420 | 118 | ||
| BC004553.2 | - | 2 - 3 | 535 | 4026156 | 118 | |
| KU178437.1 | - | 2 - 3 | 333 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000545.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
positive regulation of cytokine-mediated signaling pathway
adaptive immune response
apoptotic process
inflammatory response
signal transduction
I-kappaB kinase/NF-kappaB signaling
JNK cascade
positive regulation of peptidyl-threonine phosphorylation
peptidyl-tyrosine phosphorylation
positive regulation of protein ubiquitination
lipopolysaccharide-mediated signaling pathway
positive regulation of chemokine production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
positive regulation of interleukin-2 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of immature T cell proliferation
positive regulation of peptidyl-serine phosphorylation
toll-like receptor 2 signaling pathway
toll-like receptor 4 signaling pathway
T cell proliferation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
innate immune response
positive regulation of T-helper 1 cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of alpha-beta T cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
positive regulation of ERK1 and ERK2 cascade
nucleotide-binding oligomerization domain containing signaling pathway
nucleotide-binding oligomerization domain containing 1 signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
response to interleukin-1
response to interleukin-12
response to interleukin-18
cellular response to lipoteichoic acid
cellular response to peptidoglycan
cellular response to muramyl dipeptide
positive regulation of xenophagy
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
receptor binding
protein binding
ATP binding
LIM domain binding
protein homodimerization activity
CARD domain binding
activation of MAPK activity
positive regulation of cytokine-mediated signaling pathway
adaptive immune response
apoptotic process
inflammatory response
signal transduction
I-kappaB kinase/NF-kappaB signaling
JNK cascade
positive regulation of peptidyl-threonine phosphorylation
peptidyl-tyrosine phosphorylation
positive regulation of protein ubiquitination
lipopolysaccharide-mediated signaling pathway
positive regulation of chemokine production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
positive regulation of interleukin-2 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of immature T cell proliferation
positive regulation of peptidyl-serine phosphorylation
toll-like receptor 2 signaling pathway
toll-like receptor 4 signaling pathway
T cell proliferation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
innate immune response
positive regulation of T-helper 1 cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of alpha-beta T cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
positive regulation of ERK1 and ERK2 cascade
nucleotide-binding oligomerization domain containing signaling pathway
nucleotide-binding oligomerization domain containing 1 signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
response to interleukin-1
response to interleukin-12
response to interleukin-18
cellular response to lipoteichoic acid
cellular response to peptidoglycan
cellular response to muramyl dipeptide
positive regulation of xenophagy
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
receptor binding
protein binding
ATP binding
LIM domain binding
protein homodimerization activity
CARD domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000518673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000518673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007438.hg.1 | TC08000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007438.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007438.hg.1 | ENST00000540020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007438.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156133.hg.1 | TC0800008179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000220751.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000517696.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000518673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000518673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | ENST00000522965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008179.hg.1 | NM_003821.5 | Refseq |

RIPK2
Genomic Map