Assay Details
| Gene Symbol: MAST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527823.1 | XP_011526125.1 | 3 - 4 | 193 | 76 | |
| XM_017026511.1 | XP_016882000.1 | 2 - 3 | 491 | 76 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |

MAST3
Genomic Map