Assay Details
| Gene Symbol: LMBRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007527.1 | NP_001007528.1 | 8 - 9 | 1350 | 244 | |
| XM_011514162.2 | XP_011512464.1 | 8 - 9 | 1792 | 244 | ||
| GenBank mRNA | AK091295.1 | - | 8 - 9 | 1290 | 244 | |
| AK300433.1 | - | 9 - 10 | 1260 | 244 | ||
| AK307575.1 | - | 8 - 9 | 1296 | 244 | ||
| BX647340.1 | - | 8 - 9 | 1325 | 244 | ||
| CR749399.1 | - | 8 - 9 | 1350 | 244 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001268.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017750.hg.1 | TC05001268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017750.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017750.hg.1 | ENST00000546130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017750.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017751.hg.1 | TC05001268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017751.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017751.hg.1 | ENST00000546130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017751.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017749.hg.1 | TC05001268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017749.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017749.hg.1 | ENST00000546130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017749.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173048.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173048.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173048.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173048.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173048.hg.1 | TC0500010493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500173049.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173049.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173049.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173049.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173049.hg.1 | TC0500010493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500173047.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173047.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173047.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173047.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173047.hg.1 | TC0500010493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000296603.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000503535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | ENST00000505524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010493.hg.1 | NM_001007527.1 | Refseq |

LMBRD2
Genomic Map