Assay Details
| Gene Symbol: SPAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317931.1 | NP_001304860.1 | 5 - 6 | 685 | 72 | |
| NM_003116.2 | NP_003107.1 | 5 - 6 | 704 | 72 | ||
| XM_005260520.4 | XP_005260577.1 | 5 - 6 | 350 | 72 | ||
| XM_011529009.2 | XP_011527311.1 | 5 - 6 | 600 | 72 | ||
| GenBank mRNA | AF262992.1 | - | 5 - 6 | 701 | 72 | |
| BC143783.1 | - | 5 - 6 | 606 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000256.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20003463.hg.1 | ENST00000374273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003463.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003463.hg.1 | TC20000256.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003465.hg.1 | ENST00000374273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003465.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003465.hg.1 | TC20000256.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | TC2000007224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149761.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | TC2000007224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149763.hg.1 | NM_003116.2 | Refseq |

SPAG4
Genomic Map