Assay Details
| Gene Symbol: SPAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317931.1 | NP_001304860.1 | 7 - 8 | 822 | 75 | |
| NM_003116.2 | NP_003107.1 | 7 - 8 | 841 | 75 | ||
| XM_005260520.4 | XP_005260577.1 | 7 - 8 | 487 | 75 | ||
| XM_011529009.2 | XP_011527311.1 | 7 - 8 | 737 | 75 | ||
| GenBank mRNA | AF262992.1 | - | 7 - 8 | 838 | 75 | |
| BC143783.1 | - | 7 - 8 | 743 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000256.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20003469.hg.1 | ENST00000374273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003469.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003469.hg.1 | TC20000256.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | TC2000007224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149767.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000374273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000430878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000454819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000462896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000463973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000468248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | ENST00000498203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007224.hg.1 | NM_003116.2 | Refseq |

SPAG4
Genomic Map