Assay Details
| Gene Symbol: ARFGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006420.2 | NP_006411.2 | 20 - 21 | 2965 | 63 | |
| XM_005260252.3 | XP_005260309.1 | 20 - 21 | 2979 | 63 | ||
| GenBank mRNA | AF084521.1 | - | 20 - 21 | 2813 | 63 | |
| AK074303.1 | - | 3 - 4 | 341 | 63 | ||
| AK302526.1 | - | 6 - 7 | 820 | 63 | ||
| BC144625.1 | - | 19 - 20 | 2995 | 63 | ||
| BC144626.1 | - | 20 - 21 | 2876 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000382.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
receptor recycling
exocytosis
Golgi to plasma membrane transport
endosome organization
endomembrane system organization
protein transport
regulation of ARF protein signal transduction
positive regulation of tumor necrosis factor production
intracellular signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
ARF guanyl-nucleotide exchange factor activity
protein binding
myosin binding
protein kinase A regulatory subunit binding
GABA receptor binding
receptor recycling
exocytosis
Golgi to plasma membrane transport
endosome organization
endomembrane system organization
protein transport
regulation of ARF protein signal transduction
positive regulation of tumor necrosis factor production
intracellular signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
ARF guanyl-nucleotide exchange factor activity
protein binding
myosin binding
protein kinase A regulatory subunit binding
GABA receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20005508.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005508.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005508.hg.1 | TC20000382.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | ENST00000493140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007620.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153117.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153117.hg.1 | NM_006420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153117.hg.1 | TC2000007620.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000153117.hg.1 | ENST00000371917.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153117.hg.1 | NM_006420.2 | Refseq |

ARFGEF2
Genomic Map