Assay Details
Gene Symbol: C11orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011542639.2 | XP_011540941.1 | 2 - 3 | 113 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011542639.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000393084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000524755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000525729.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000526725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000527531.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000529391.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000533583.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | ENST00000615746.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012227.hg.1 | NM_152587.4 | Refseq |
