Assay Details
| Gene Symbol: ITPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002224.3 | NP_002215.2 | 26 - 27 | 3676 | 59 | |
| XM_011514577.2 | XP_011512879.1 | 25 - 26 | 3316 | 59 | ||
| XM_017010832.1 | XP_016866321.1 | 27 - 28 | 3505 | 59 | ||
| GenBank mRNA | AB209739.1 | - | 17 - 18 | 2463 | 59 | |
| D26351.1 | - | 26 - 27 | 3687 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000427.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
memory
platelet activation
inositol phosphate-mediated signaling
regulation of insulin secretion
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
release of sequestered calcium ion into cytosol
protein homooligomerization
protein heterooligomerization
response to calcium ion
long-term synaptic potentiation
calcium ion transport into cytosol
cellular response to cAMP
regulation of cardiac conduction
inositol hexakisphosphate binding
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium ion binding
protein binding
calcium-release channel activity
phosphatidylinositol binding
inositol 1,3,4,5 tetrakisphosphate binding
inositol 1,4,5 trisphosphate binding
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
memory
platelet activation
inositol phosphate-mediated signaling
regulation of insulin secretion
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
release of sequestered calcium ion into cytosol
protein homooligomerization
protein heterooligomerization
response to calcium ion
long-term synaptic potentiation
calcium ion transport into cytosol
cellular response to cAMP
regulation of cardiac conduction
inositol hexakisphosphate binding
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium ion binding
protein binding
calcium-release channel activity
phosphatidylinositol binding
inositol 1,3,4,5 tetrakisphosphate binding
inositol 1,4,5 trisphosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000374316.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | ENST00000605930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007711.hg.1 | NM_002224.3 | Refseq |

ITPR3
Genomic Map