Assay Details
| Gene Symbol: ITPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002224.3 | NP_002215.2 | 34 - 35 | 4824 | 68 | |
| XM_011514577.2 | XP_011512879.1 | 33 - 34 | 4464 | 68 | ||
| GenBank mRNA | AB209739.1 | - | 25 - 26 | 3611 | 68 | |
| D26351.1 | - | 34 - 35 | 4835 | 68 | ||
| U01062.1 | - | 34 - 35 | 4636 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000427.hg.1
Set Membership: |
|

ITPR3
Genomic Map