Assay Details
| Gene Symbol: PPM1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538578.2 | XP_011536880.1 | 1 - 2 | 532 | 64 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000228705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000547857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000548414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000551519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | ENST00000619248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011054.hg.1 | NM_020700.1 | Refseq |

PPM1H
Genomic Map