Assay Details
| Gene Symbol: SLC5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000343.3 | NP_000334.1 | 13 - 14 | 1912 | 58 | |
| NM_001256314.1 | NP_001243243.1 | 12 - 13 | 1630 | 58 | ||
| GenBank mRNA | AK297665.1 | - | 12 - 13 | 1604 | 58 | |
| AK312948.1 | - | 13 - 14 | 1816 | 58 | ||
| AK316184.1 | - | 12 - 13 | 1609 | 58 | ||
| BC139839.1 | - | 13 - 14 | 1681 | 58 | ||
| CR456579.1 | - | 13 - 14 | 1712 | 58 | ||
| M24847.1 | - | 13 - 14 | 1672 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000237.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22004576.hg.1 | ENST00000266088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004576.hg.1 | ENST00000543737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004576.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004576.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004576.hg.1 | TC22000237.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004577.hg.1 | ENST00000266088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004577.hg.1 | ENST00000543737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004577.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004577.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004577.hg.1 | TC22000237.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000477969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000486394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007120.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | TC2200007120.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150183.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | TC2200007120.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | NM_001256314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | ENST00000266088.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | ENST00000543737.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | NM_000343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150184.hg.1 | NM_001256314.1 | Refseq |

SLC5A1
Genomic Map