Assay Details
| Gene Symbol: TSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046308.1 | 1 - 2 | 96 | 95 | ||
| GenBank mRNA | L21950.1 | - | 1 - 2 | 66 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the L21950.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to mitochondrion
steroid biosynthetic process
heme biosynthetic process
anion transport
chloride transport
lipid transport
apoptotic process
signal transduction
chemical synaptic transmission
aging
steroid metabolic process
cell proliferation
glial cell migration
response to manganese ion
response to vitamin B1
positive regulation of necrotic cell death
peripheral nervous system axon regeneration
adrenal gland development
negative regulation of protein ubiquitination
regulation of cholesterol transport
response to progesterone
negative regulation of tumor necrosis factor production
response to testosterone
response to drug
positive regulation of apoptotic process
negative regulation of nitric oxide biosynthetic process
behavioral response to pain
regulation of steroid biosynthetic process
positive regulation of mitochondrial depolarization
positive regulation of calcium ion transport
contact inhibition
positive regulation of glial cell proliferation
negative regulation of glial cell proliferation
cellular response to lipopolysaccharide
cellular response to zinc ion
cellular hypotonic response
maintenance of protein location in mitochondrion
negative regulation of mitophagy
negative regulation of ATP metabolic process
positive regulation of reactive oxygen species metabolic process
androgen binding
protein binding
benzodiazepine receptor activity
cholesterol binding
ion channel binding
protein targeting to mitochondrion
steroid biosynthetic process
heme biosynthetic process
anion transport
chloride transport
lipid transport
apoptotic process
signal transduction
chemical synaptic transmission
aging
steroid metabolic process
cell proliferation
glial cell migration
response to manganese ion
response to vitamin B1
positive regulation of necrotic cell death
peripheral nervous system axon regeneration
adrenal gland development
negative regulation of protein ubiquitination
regulation of cholesterol transport
response to progesterone
negative regulation of tumor necrosis factor production
response to testosterone
response to drug
positive regulation of apoptotic process
negative regulation of nitric oxide biosynthetic process
behavioral response to pain
regulation of steroid biosynthetic process
positive regulation of mitochondrial depolarization
positive regulation of calcium ion transport
contact inhibition
positive regulation of glial cell proliferation
negative regulation of glial cell proliferation
cellular response to lipopolysaccharide
cellular response to zinc ion
cellular hypotonic response
maintenance of protein location in mitochondrion
negative regulation of mitophagy
negative regulation of ATP metabolic process
positive regulation of reactive oxygen species metabolic process
androgen binding
protein binding
benzodiazepine receptor activity
cholesterol binding
ion channel binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000329563.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000396265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000428336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NM_001256531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007547.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | TC2200007547.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | NR_046308.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000337554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000472378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | ENST00000583777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | NM_000714.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200153513.hg.1 | NR_046308.1 | Refseq |

TSPO
Genomic Map