Assay Details
| Gene Symbol: GPR78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080819.4 | NP_543009.2 | 1 - 2 | 1156 | 87 | |
| GenBank mRNA | AF411107.1 | - | 1 - 2 | 663 | 87 | |
| AK128807.1 | - | 1 - 2 | 1156 | 87 | ||
| AK313038.1 | - | 1 - 2 | 731 | 87 | ||
| AY359107.1 | - | 1 - 2 | 957 | 87 | ||
| BC057778.1 | - | 1 - 2 | 1080 | 30343110 | 87 | |
| BC069343.1 | - | 1 - 2 | 663 | 7262307 | 87 | |
| BC069813.1 | - | 1 - 2 | 663 | 7262319 | 87 | |
| JF810885.1 | - | 1 - 2 | 663 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002932.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04001919.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001919.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001919.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001919.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001923.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001923.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001923.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001920.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001920.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001920.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001920.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001924.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001924.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001924.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001925.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001925.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001925.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001925.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001925.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001921.hg.1 | TC04002932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001921.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001921.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001921.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001921.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187803.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187803.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187803.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187803.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187803.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187801.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187799.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400187804.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187804.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187804.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187804.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187804.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187805.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400187800.hg.1 | TC0400012764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000382487.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000503981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000504255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | ENST00000509216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NM_080819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012764.hg.1 | NR_045511.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 117667 | - | - |

GPR78
Genomic Map