Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289.5 | NP_001280.3 | 1 - 2 | 315 | 72 | |
| GenBank mRNA | AK292785.1 | - | 1 - 2 | 248 | 72 | |
| BC005367.1 | - | 1 - 2 | 191 | 72 | ||
| BC022305.1 | - | 1 - 2 | 191 | 4663624 | 72 | |
| BX482109.1 | - | 1 - 2 | 310 | 72 | ||
| DA901964.1 | - | 1 - 2 | 315 | 72 | ||
| Y12696.1 | - | 1 - 2 | 282 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001551.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
transport
signal transduction
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of binding
negative regulation of ryanodine-sensitive calcium-release channel activity
cellular oxidant detoxification
chloride transmembrane transport
glutathione transferase activity
glutathione peroxidase activity
voltage-gated ion channel activity
chloride channel activity
protein binding
glutathione metabolic process
transport
signal transduction
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of binding
negative regulation of ryanodine-sensitive calcium-release channel activity
cellular oxidant detoxification
chloride transmembrane transport
glutathione transferase activity
glutathione peroxidase activity
voltage-gated ion channel activity
chloride channel activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020812.hg.1 | TC0X001551.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020812.hg.1 | ENST00000369449.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020812.hg.1 | ENST00000465553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020812.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020813.hg.1 | TC0X001551.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020813.hg.1 | ENST00000369449.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020813.hg.1 | ENST00000465553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020813.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020810.hg.1 | TC0X001551.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020810.hg.1 | ENST00000369449.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020810.hg.1 | ENST00000465553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020810.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020815.hg.1 | TC0X001551.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020815.hg.1 | ENST00000369449.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020815.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176889.hg.1 | TC0X00011238.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176886.hg.1 | TC0X00011238.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011238.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000465553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | ENST00000491205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176888.hg.1 | TC0X00011238.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | ENST00000321926.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | ENST00000369449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | NM_001289.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176891.hg.1 | TC0X00011238.hg.1 | Clariom D TC |

CLIC2
Genomic Map