Assay Details
| Gene Symbol: PPP6R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242899.1 | NP_001229828.1 | 15 - 16 | 2080 | 137 | |
| NM_001242900.1 | NP_001229829.1 | 15 - 16 | 2077 | 137 | ||
| NM_014678.4 | NP_055493.2 | 15 - 16 | 2077 | 137 | ||
| XM_006724435.2 | XP_006724498.1 | 15 - 16 | 2062 | 137 | ||
| XM_011530726.2 | XP_011529028.1 | 14 - 15 | 1765 | 137 | ||
| XM_011530727.2 | XP_011529029.1 | 14 - 15 | 1765 | 137 | ||
| XM_011530728.2 | XP_011529030.1 | 14 - 15 | 1765 | 137 | ||
| XM_011530729.2 | XP_011529031.1 | 14 - 15 | 1765 | 137 | ||
| XM_011530730.2 | XP_011529032.1 | 14 - 15 | 1762 | 137 | ||
| XM_011530731.1 | XP_011529033.1 | 14 - 15 | 1762 | 137 | ||
| XM_017029116.1 | XP_016884605.1 | 14 - 15 | 1762 | 137 | ||
| XM_017029129.1 | XP_016884618.1 | 15 - 16 | 2062 | 137 | ||
| XM_017029130.1 | XP_016884619.1 | 15 - 16 | 2062 | 137 | ||
| XM_017029131.1 | XP_016884620.1 | 16 - 17 | 2188 | 137 | ||
| XM_017029132.1 | XP_016884621.1 | 16 - 17 | 2140 | 137 | ||
| XM_017029138.1 | XP_016884627.1 | 15 - 16 | 2016 | 137 | ||
| GenBank mRNA | AB014585.1 | - | 15 - 16 | 2049 | 137 | |
| BC006568.2 | - | 15 - 16 | 2063 | 3532211 | 137 | |
| BC032664.1 | - | 15 - 16 | 2077 | 5553142 | 137 | |
| BC041698.1 | - | 15 - 16 | 2047 | 5732101 | 137 | |
| BC052995.2 | - | 15 - 16 | 1983 | 6162781 | 137 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PPP6R2
Genomic Map