| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015662.hg.1 |
TC0X001162.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015662.hg.1 |
ENST00000332687.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015662.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015662.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015662.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
ENST00000469174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168888.hg.1 |
TC0X00010097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000332687.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000349225.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
ENST00000469174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_016120.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010097.hg.1 |
NM_183353.2 |
Refseq |