Assay Details
| Gene Symbol: MSANTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042690.1 | NP_001036155.1 | 1 - 2 | 503 | 116 | |
| XM_011513465.2 | XP_011511767.1 | 4 - 5 | 1765 | 116 | ||
| XM_011513466.2 | XP_011511768.1 | 2 - 3 | 4030 | 116 | ||
| XM_017008169.1 | XP_016863658.1 | 2 - 3 | 3208 | 116 | ||
| GenBank mRNA | AK126094.1 | - | 1 - 2 | 503 | 116 | |
| BC043530.1 | - | 4 - 5 | 1124 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000045.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001202.hg.1 | TC04000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001202.hg.1 | ENST00000438480.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001202.hg.1 | ENST00000507492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001202.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001202.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001199.hg.1 | TC04000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001199.hg.1 | ENST00000438480.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001199.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001200.hg.1 | TC04000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001200.hg.1 | ENST00000438480.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001200.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001200.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001201.hg.1 | TC04000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001201.hg.1 | ENST00000438480.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001201.hg.1 | ENST00000507492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001201.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001201.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145666.hg.1 | TC0400006596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145665.hg.1 | TC0400006596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145668.hg.1 | TC0400006596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145669.hg.1 | TC0400006596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145667.hg.1 | TC0400006596.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000438480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000505599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000507492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | ENST00000510580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006596.hg.1 | NM_001042690.1 | Refseq |

MSANTD1
Genomic Map