Assay Details
| Gene Symbol: CPSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322270.1 | NP_001309199.1 | 3 - 4 | 412 | 158 | |
| NM_001322272.1 | NP_001309201.1 | 3 - 4 | 617 | 158 | ||
| NM_017437.2 | NP_059133.1 | 3 - 4 | 412 | 158 | ||
| GenBank mRNA | AK001627.2 | - | 3 - 4 | 389 | 158 | |
| AK001912.1 | - | 3 - 4 | 382 | 158 | ||
| AU132572.1 | - | 3 - 4 | 389 | 158 | ||
| BC070095.1 | - | 3 - 4 | 382 | 5267724 | 158 | |
| BU071571.1 | - | 1 - 2 | 63 | 158 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000568.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14006894.hg.1 | ENST00000298875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006894.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006894.hg.1 | TC14000568.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156625.hg.1 | TC1400008007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000298875.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000553427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000554290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000555244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | ENST00000556622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008007.hg.1 | NM_017437.2 | Refseq |

CPSF2
Genomic Map